SY3H3D6755CA Synthonix Corporation

(2-Aminophenyl)(4-fluorophenyl)methanone

  • MDL number MFCD06658166
  • Cas Number : 3800-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39694.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39694.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart