SY3H3D6755CD Synthonix Corporation

(Z)-2-(2-Aminothiazol-4-yl)-2-(((1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl)oxy)imino)acetic acid

  • MDL number MFCD00071569
  • Cas Number : 86299-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39709.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39709.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart