SY3H3D6755DD Synthonix Corporation

3-Amino-1H-pyrazol-5(4H)-one

  • MDL number MFCD01084003
  • Cas Number : 6126-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Greyish yellow red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39854.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39854.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart