SY3H3D6755DF Synthonix Corporation

5-Amino-3-methyl-1,2,4-thiadiazole

  • MDL number MFCD01114751
  • Cas Number : 17467-35-5
  • Molecular Weight : 115.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39858.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39858.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart