SY3H3D6755EE Synthonix Corporation

2-Cyanobenzyl acetate

  • MDL number MFCD20265196
  • Cas Number : 38866-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/0595.html

Safety Information

MSDS https://synthonix.com/0595.html
Availability details for
Quantity
  • Availability
  • Add to Cart