SY3H3D6755EF Synthonix Corporation

4-Amino-1-methyl-4H-1,2,4-triazol-1-ium nitrate

  • MDL number MFCD28053750
  • Cas Number : 817177-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39940.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39940.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart