SY3H3D6756D9 Synthonix Corporation

5-Aminoisoxazole-4-carbonitrile

  • MDL number MFCD00128284
  • Cas Number : 98027-17-9
  • Molecular Weight : 109.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40053.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40053.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart