SY3H3D6756E2 Synthonix Corporation

6-Amino-5-bromopyrimidin-4(3H)-one

  • MDL number MFCD08234407
  • Cas Number : 6312-71-6
  • Molecular Weight : 190

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40094.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40094.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart