SY3H3D6756E3 Synthonix Corporation

2-Amino-5-bromopyrimidin-4-ol hydrobromide

  • MDL number MFCD12923111
  • Cas Number : 1215597-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40095.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40095.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart