SY3H3D6756EA Synthonix Corporation

6-Amino-5-nitropyrimidine-2,4(1H,3H)-dione

  • MDL number No MDL found
  • Cas Number : 3346-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/40103.html

Safety Information

MSDS https://synthonix.com/40103.html
Availability details for
Quantity
  • Availability
  • Add to Cart