SY3H3D6756FD Synthonix Corporation

2-Amino-N,N-dimethylacetamide hydrochloride

  • MDL number MFCD08060117
  • Cas Number : 72287-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to Yellow Solid or Semi-solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40307.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40307.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart