SY3H3D67570F Synthonix Corporation

(R)-2-Amino-3-methoxypropanoic acid hydrochloride

  • MDL number MFCD11099371
  • Cas Number : 86118-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40353.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40353.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart