SY3H3D675729 Synthonix Corporation

2-Amino-4-hydroxythiophene-3-carbonitrile

  • MDL number No MDL found
  • Cas Number : 99580-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40780.html

Safety Information

MSDS https://synthonix.com/40780.html
Availability details for
Quantity
  • Availability
  • Add to Cart