SY3H3D675768 Synthonix Corporation

6-Amino-3-methylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD02091062
  • Cas Number : 21236-97-5
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41149.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/41149.html
pictograms GHS07
hstatements H302, H312, H332
Availability details for
Quantity
  • Availability
  • Add to Cart