SY3H3D675773 Synthonix Corporation

2-Amino-4,6-dimethoxy-1,3,5-triazine

  • MDL number MFCD00052765
  • Cas Number : 16370-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41214.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41214.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart