SY3H3D675786 Synthonix Corporation

(R)-2-Aminopent-4-ynoic acid hydrochloride

  • MDL number MFCD09263338
  • Cas Number : 87205-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41354.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41354.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart