SY3H3D6757A6 Synthonix Corporation

3-Amino-2-bromo-6-(trifluoromethyl)pyridine

  • MDL number MFCD11052334
  • Cas Number : 117519-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42070.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42070.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart