SY3H3D6757B9 Synthonix Corporation

2-Amino-4-bromo-6-nitrophenol

  • MDL number MFCD22206403
  • Cas Number : 139138-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42429.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42429.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart