SY3H3D6757D7 Synthonix Corporation

3-Acetyl-5-chlorothiophene-2-sulfonamide

  • MDL number MFCD09033309
  • Cas Number : 160982-10-5
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42871.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42871.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart