SY3H3D6757DA Synthonix Corporation

4-Amino-2-bromophenol

  • MDL number MFCD06656565
  • Cas Number : 16750-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Pale gray - Reddish gray - Black
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42878.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42878.html
pictograms GHS07
hstatements H302, H315, H319
Availability details for
Quantity
  • Availability
  • Add to Cart