SY3H3D6757E8 Synthonix Corporation

5-Amino-2-bromo-3-picoline

  • MDL number MFCD03095085
  • Cas Number : 38186-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Pale yellow - Greyish yellow red - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42967.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42967.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart