SY3H3D6757E9 Synthonix Corporation

3-Amino-2-bromo-5-methylpyridine

  • MDL number MFCD08277281
  • Cas Number : 34552-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42977.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42977.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart