SY3H3D6757F9 Synthonix Corporation

6-Amino-3-nitro-2-picoline

  • MDL number MFCD00053582
  • Cas Number : 22280-62-2
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Gray - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43103.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43103.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart