SY3H3D675803 Synthonix Corporation

2-Aminocyclopent-1-enecarbonitrile

  • MDL number MFCD00517551
  • Cas Number : 2941-23-3
  • Molecular Weight : 108.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43185.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43185.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart