SY3H3D67580A Synthonix Corporation

4-Amino-5-methylpyridin-2-ol

  • MDL number MFCD12546373
  • Cas Number : 95306-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43204.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43204.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart