SY3H3D67581D Synthonix Corporation

(R)-3-Acetylthiazolidine-4-carboxylic acid

  • MDL number MFCD00038440
  • Cas Number : 54323-50-1
  • Molecular Weight : 175.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43391.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43391.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart