SY3H3D675824 Synthonix Corporation

(1S,3S)-3-Aminocyclopentanecarboxylic acid hydrochloride

  • MDL number MFCD30181867
  • Cas Number : 2243913-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43450.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43450.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart