SY3H3D67582C Synthonix Corporation

3-Aminocyclohexanol hydrochloride

  • MDL number MFCD18432505
  • Cas Number : 1263378-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43477.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43477.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart