SY3H3D675830 Synthonix Corporation

(1S,2S)-2-Aminocyclohexanol hydrochloride

  • MDL number MFCD09259963
  • Cas Number : 13374-30-6
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form crystalline Colour: white to yellow to light-brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43482.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43482.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart