SY3H3D67588E Synthonix Corporation

5-Amino-2-chlorobenzothiazole

  • MDL number MFCD11040282
  • Cas Number : 80945-82-0
  • Molecular Weight : 184.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44787.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44787.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart