SY3H3D6758A0 Synthonix Corporation

3-Amino-5-fluorobenzotrifluoride

  • MDL number MFCD00061315
  • Cas Number : 454-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44920.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44920.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart