SY3H3D6758B0 Synthonix Corporation

3-Amino-2,6-difluorobenzoic acid

  • MDL number MFCD07368833
  • Cas Number : 83141-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Light-red to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44988.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44988.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart