SY3H3D6758C0 Synthonix Corporation

2-Amino-4-nitrobenzaldehyde

  • MDL number MFCD09264065
  • Cas Number : 109466-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45244.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45244.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart