SY3H3D6758C4 Synthonix Corporation

2-Amino-6-nitrobenzoic acid

  • MDL number MFCD00034785
  • Cas Number : 50573-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Pale yellow - Yellow red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45253.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45253.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart