SY3H3D6758ED Synthonix Corporation

2-Amino-5-chlorobenzamide

  • MDL number MFCD00017126
  • Cas Number : 5202-85-7
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/45696.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45696.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart