SY3H3D67590D Synthonix Corporation

(R)-2-Amino-3-(5-bromothiophen-2-yl)propanoic acid

  • MDL number MFCD04117831
  • Cas Number : 264903-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46137.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46137.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart