SY3H3D67590E Synthonix Corporation

(S)-2-Amino-3-(5-bromothiophen-2-yl)propanoic acid

  • MDL number MFCD04117834
  • Cas Number : 154593-58-5
  • Molecular Weight : 250.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46138.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46138.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart