SY3H3D675913 Synthonix Corporation

(2-Amino-4-bromophenyl)methanol

  • MDL number No MDL found
  • Cas Number : 946122-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46164.html

Safety Information

MSDS https://synthonix.com/46164.html
Availability details for
Quantity
  • Availability
  • Add to Cart