SY3H3D675918 Synthonix Corporation

(2-Amino-5-chlorophenyl)methanol

  • MDL number MFCD00075163
  • Cas Number : 37585-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46196.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46196.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart