SY3H3D67591F Synthonix Corporation

(3-Amino-2-fluorophenyl)methanol

  • MDL number MFCD11846488
  • Cas Number : 1051899-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light yellow to yellow to yellow-brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46217.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46217.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart