SY3H3D675929 Synthonix Corporation

2-(Azetidin-3-yl)pyrazine

  • MDL number MFCD19227705
  • Cas Number : 1236861-70-3
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46337.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46337.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart