SY3H3D67592D Synthonix Corporation

N-(6-Aminopyridin-2-yl)acetamide

  • MDL number MFCD01099052
  • Cas Number : 1075-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46342.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/46342.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart