SY3H3D675932 Synthonix Corporation

4-Amino-2-methylphenol

  • MDL number No MDL found
  • Cas Number : 2835-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46374.html

Safety Information

MSDS https://synthonix.com/46374.html
Availability details for
Quantity
  • Availability
  • Add to Cart