SY3H3D67593A Synthonix Corporation

(R)-2-Amino-3-(thiophen-2-yl)propanoic acid

  • MDL number MFCD00077166
  • Cas Number : 62561-76-6
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46420.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46420.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart