SY3H3D67594C Synthonix Corporation

(S)-2-Amino-3-(tert-butylthio)propanoic acid hydrochloride

  • MDL number MFCD00135154
  • Cas Number : 200353-65-7
  • Molecular Weight : 213.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46521.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46521.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart