SY3H3D675983 Synthonix Corporation

4-Amino-3,5-dimethylphenol

  • MDL number MFCD01741459
  • Cas Number : 3096-70-6
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/46903.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/46903.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart