SY3H3D675984 Synthonix Corporation

3-(Aminomethyl)benzyl Alcohol

  • MDL number No MDL found
  • Cas Number : 34231-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46908.html

Safety Information

MSDS https://synthonix.com/46908.html
Availability details for
Quantity
  • Availability
  • Add to Cart