SY3H3D67598A Synthonix Corporation

(2-Amino-5-methoxyphenyl)methanol

  • MDL number MFCD09966091
  • Cas Number : 55414-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to Light-gray to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46923.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46923.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart