SY3H3D67599B Synthonix Corporation

(1-Allylpyrrolidin-2-yl)methanamine

  • MDL number MFCD01317559
  • Cas Number : 26116-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47145.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47145.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart