SY3H3D6759A6 Synthonix Corporation

2-Amino-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD03407465

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to Yellow to Brown Solid or Semi-Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47559.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47559.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart